• USA Home
  • 100439 - Acetylcholine iodide

EMAIL THIS PAGE TO A FRIEND
100439 Aldrich

Acetylcholine iodide

98%

Synonym: ACh

Purchase

Properties

InChI Key   SMBBQHHYSLHDHF-UHFFFAOYSA-M
assay   98%

Description

Biochem/physiol Actions

Endogenous neurotransmitter at cholinergic synapses; amplifies action potential of the sarcolemma thereby inducing muscle contractions.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
KH3300000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?