• USA Home
  • 100633 - 1-(2-Hydroxyphenyl)-3-phenyl-2-propenone

EMAIL THIS PAGE TO A FRIEND
100633 Aldrich

1-(2-Hydroxyphenyl)-3-phenyl-2-propenone

98%

Synonym: 2′-Hydroxychalcone, 2-Hydroxybenzalacetophenone

  • CAS Number 1214-47-7

  • Linear Formula C6H5CH=CHCOC6H4OH

  • Molecular Weight 224.25

  •  Beilstein Registry Number 976324

  •  EC Number 214-928-0

  •  MDL number MFCD00016441

  •  PubChem Substance ID 24846420

Purchase

Properties

Related Categories Building Blocks, C15 to C38, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   AETKQQBRKSELEL-ZHACJKMWSA-N
assay   98%
mp   86-88 °C(lit.)
Gene Information   human ... IL1B(3553)
rat ... Alox5(25290)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UD5579400

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?