• USA Home
  • 105112 - Chloromethyl thiocyanate

EMAIL THIS PAGE TO A FRIEND
105112 Aldrich

Chloromethyl thiocyanate

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocyanates/Isothiocyanates More...
InChI Key   UXUCVNXUWOLPRU-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.515(lit.)
bp   76-78 °C/15 mmHg(lit.)
density   1.37 g/mL at 25 °C(lit.)

Description

Packaging

10 g in glass bottle

General description

Chloromethyl thiocyanate reacts with 1-methylimidazole to form imidazolium salt, 1-thiocyanomethyl-3-methylimidazolium chloride.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN 2810 6.1 / PGII
WGK Germany 
3
RTECS 
XK8800000
Flash Point(F) 
183.2 °F
Flash Point(C) 
84 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?