EMAIL THIS PAGE TO A FRIEND
10615 Aldrich

Holmium(III) oxide

puriss., ≥99.9%, powder

Synonym: Rare earth

Purchase

Properties

InChI Key   JYTUFVYWTIKZGR-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.9%
form   powder
mp   2330 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?