EMAIL THIS PAGE TO A FRIEND
107883 Aldrich

Methyl benzilate

99%

Synonym: Methyl diphenylglycolate

  • CAS Number 76-89-1

  • Linear Formula (C6H5)2C(OH)COOCH3

  • Molecular Weight 242.27

  •  Beilstein Registry Number 2121492

  •  MDL number MFCD00004446

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   LJFIHTFNTGQZJL-UHFFFAOYSA-N
assay   99%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?