• USA Home
  • 10883 - 1,4-Dithiane-2,5-diol

EMAIL THIS PAGE TO A FRIEND
10883 Aldrich

1,4-Dithiane-2,5-diol

produced by Wacker Chemie AG, Burghausen, Germany, ≥96% (GC)

Synonym: 2,5-Dihydroxy-1,4-dithiane, Dithian, Mercaptoacetaldehyde dimer

  • CAS Number 40018-26-6

  • Empirical Formula (Hill Notation) C4H8O2S2

  • Molecular Weight 152.24

  •  Beilstein Registry Number 104402

  •  EC Number 254-751-6

  •  MDL number MFCD00006659

  •  PubChem Substance ID 57646826

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Other Heterocyclic Building Blocks, S-Containing More...
grade   produced by Wacker Chemie AG, Burghausen, Germany
InChI Key   YUIOPHXTILULQC-UHFFFAOYSA-N
assay   ≥96% (GC)
mfr. no.   Wacker Chemie AG
mp   130 °C (dec.)(lit.)
solubility   H2O: soluble0.39% at 20 °C, clear, colorless
storage temp.   2-8°C

Description

Other Notes

prices for bulk quantities on request

Packaging

5 kg in poly bottle

Application

1,4-Dithiane-2,5-diol was used to analyze the different aroma volatiles of the cooked meat from animals fed dietary polyunsaturated fatty acids supplements.

General description

1,4-Dithiane-2,5-diol is the dimer of mercaptoacetaldehyde. It is a suitable bifunctional reagent. It undergoes Michael intermolecular addition to in situ generated nitroalkenes to yield 3,4-substituted tetrahydrothiophenes.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3335
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?