• USA Home
  • 10909 - N,N-Dimethylacetoacetamide solution

EMAIL THIS PAGE TO A FRIEND
10909 Aldrich

N,N-Dimethylacetoacetamide solution

produced by Wacker Chemie AG, Burghausen, Germany, 80% in H2O (GC)

Synonym: N,N-Dimethyl-3-oxobutyramide, DMAA

  • CAS Number 2044-64-6

  • Linear Formula CH3COCH2CON(CH3)2

  • Molecular Weight 129.16

  •  Beilstein Registry Number 1755038

  •  MDL number MFCD00038243

  •  PubChem Substance ID 57646843

Purchase

Properties

Related Categories Amides, Building Blocks, C2 to C7, Carbonyl Compounds, Chemical Synthesis,
grade   produced by Wacker Chemie AG, Burghausen, Germany
InChI Key   YPEWWOUWRRQBAX-UHFFFAOYSA-N
mfr. no.   Wacker Chemie AG
concentration   80% in H2O (GC)
solubility   H2O: soluble

Description

Other Notes

prices for bulk quantities on request

General description

N,N-Dimethylacetoacetamide on mixing with HAuCl4 forms uniform Au nanoparticles. It is the major hydrolytic degradation product of dicrotophos, an active ingredient of Bidrin insecticide.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
Flash Point(F) 
251.6 °F
Flash Point(C) 
122 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?