10917 Aldrich

Dibutyl ether

produced by Wacker, ≥99.3% (GC)

DOWNLOAD MSDS (PDF)

Synonym: Butyl ether, DBE

  • CAS Number 142-96-1

  • Linear Formula [CH3(CH2)3]2O

  • Molecular Weight 130.23

  •  Beilstein Registry Number 1732752

  •  EC Number 205-575-3

  •  MDL number MFCD00009461

  •  PubChem Substance ID 24846891

Properties

Related Categories Building Blocks, C8, Chemical Synthesis, Ethers, Organic Building Blocks,
grade   produced by Wacker
vapor density   4.48 (vs air)
vapor pressure   4.8 mmHg ( 20 °C)
assay   ≥99.3% (GC)
autoignition temp.   365 °F
expl. lim.   8.5 %
mfr. no.   Wacker Chemie AG
refractive index   n20/D 1.399(lit.)
bp   142-143 °C(lit.)
mp   −98 °C(lit.)
density   0.764 g/mL at 25 °C(lit.)

Description

Other Notes

prices for bulk quantities on request

Price and Availability

Customers Also Viewed

Dibutyl ether

anhydrous, 99.3%

Dibutyl ether

produced by Wacker, ≥99.3% (GC)

Dibutyl ether

puriss. p.a., low in aromatic compounds, ≥99.5% (GC)

Dibutyl ether

purum, ≥99.0% (GC)

Dibutyl ether

ReagentPlus®, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?