• USA Home
  • 110310 - Diethylene glycol butyl ether

EMAIL THIS PAGE TO A FRIEND
110310 Aldrich

Diethylene glycol butyl ether

≥99%

Synonym: 2-(2-Butoxyethoxy)ethanol, BDG, Butyldiglycol

  • CAS Number 112-34-5

  • Linear Formula CH3(CH2)3OCH2CH2OCH2CH2OH

  • Molecular Weight 162.23

  •  Beilstein Registry Number 1739225

  •  EC Number 203-961-6

  •  MDL number MFCD00002881

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

vapor density   5.6 (vs air)
vapor pressure   30 mmHg ( 130 °C)
InChI Key   OAYXUHPQHDHDDZ-UHFFFAOYSA-N
assay   ≥99%
autoignition temp.   400 °F
expl. lim.   6.2 %
refractive index   n20/D 1.432(lit.)
bp   231 °C(lit.)
  231 °C
mp   -68 °C(lit.)
  −68 °C(lit.)
density   0.967 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
KJ9100000
Flash Point(F) 
210.2 °F
Flash Point(C) 
99 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?