• USA Home
  • 112410 - Dimethylnaphthalene, mixture of isomers

EMAIL THIS PAGE TO A FRIEND
112410 Aldrich

Dimethylnaphthalene, mixture of isomers

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Organic Building Blocks
InChI Key   YGYNBBAUIYTWBF-UHFFFAOYSA-N
refractive index   n20/D 1.6088(lit.)
density   1.01 g/mL at 25 °C(lit.)
Gene Information   human ... CYP1A2(1544), CYP2A6(1548)
mouse ... Cyp2a5(13087)

Description

Application

Pollutant / Analyte involved in:
• Reactions with chlorine atoms for assessing the reactions environmental impact
• Studies acting as a marker ion for heavy-duty vehicle emissions and investigations into stability of single particle tracers
• Effect of pretreatment with carbonic acid on "HyperCoal" or ash-free coal production
• Comparing the response of biomarkers to diagnose ecological impact of iol spills
• Investigating the effects of polymer modification on the properties and microstructure of model asphalt systems
• Applying Hansen solubility parameters theory to carbon nanotubes

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?