EMAIL THIS PAGE TO A FRIEND
115932 Aldrich

6-Nitrophthalide

95%

  • CAS Number 610-93-5

  • Empirical Formula (Hill Notation) C8H5NO4

  • Molecular Weight 179.13

  •  MDL number MFCD00033529

  •  PubChem Substance ID 24847252

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Chemical Synthesis, Lactones, Organic Building Blocks More...
InChI Key   RNWGZXAHUPFXLL-UHFFFAOYSA-N
assay   95%
mp   140-145 °C(lit.)

Description

Packaging

10 g in glass bottle

Application

6-Nitrophthalide has been used in the preparation of 3-bromo-6-nitrophthalide. It undergoes reductive methylation to form 6-dimethylaminophthalide.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?