• USA Home
  • 11846 - (+)-Barium tartrate dibasic

EMAIL THIS PAGE TO A FRIEND
11846 Aldrich

(+)-Barium tartrate dibasic

purum, ≥98.0%

Synonym: L-(+)-Tartaric acid barium salt

  • CAS Number 5908-81-6

  • Linear Formula OOCCH(OH)CH(OH)COOBa

  • Molecular Weight 285.40

  •  Beilstein Registry Number 6120238

  •  EC Number 227-615-9

  •  MDL number MFCD00054437

  •  PubChem Substance ID 57646957

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acid Salts, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   HQYOWPDUMCBSLQ-UHFFFAOYSA-L
grade   purum
assay   ≥98.0%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1564 6.1/PG 3
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?