• USA Home
  • 11901 - Bromide on polymer support
EMAIL THIS PAGE TO A FRIEND
11901 Aldrich
Bromide on polymer support
extent of labeling: ~3.5 mmol/g loading
Purchase
Properties
Related Categories Chemical Synthesis, Other Resins, Solid Supported Synthesis, Specialty Synthesis
extent of labeling   ~3.5 mmol/g loading
Description
Other Notes
Polymer supported reagent for the preparation of a alkyl bromides from alkyl chlorides
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?