EMAIL THIS PAGE TO A FRIEND
119962 Aldrich

Alizarin Red S

certified by the Biological Stain Commission, Dye content 70 %

Synonym: 3,4-Dihydroxy-9,10-dioxo-2-anthracenesulfonic acid sodium salt, Alizarin Carmine, Alizarin Red, water soluble, Alizarin sodium monosulfonate, Alizarin sulfonate sodium, Alizarinsulfonic acid sodium salt, Sodium alizarinsulfonate

  • CAS Number 130-22-3

  • Empirical Formula (Hill Notation) C14H7NaO7S

  • Molecular Weight 342.26

  •  Beilstein Registry Number 4117090

  •  Colour Index Number 58005

  •  EC Number 204-981-8

  •  MDL number MFCD00013049

Purchase

Properties

grade   certified by the Biological Stain Commission
InChI Key   HFVAFDPGUJEFBQ-UHFFFAOYSA-M
composition   Dye content, 70%
absorption   λmax 556 nm
  λmax 596 nm

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

A5533 Alizarin Red S, certified by the Biological Stain Commission

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?