• USA Home
  • 121444 - 1,2-Di-p-tolylethane

EMAIL THIS PAGE TO A FRIEND
121444 Aldrich

1,2-Di-p-tolylethane

97%

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Organic Building Blocks
InChI Key   XCCQFUHBIRHLQT-UHFFFAOYSA-N
assay   97%
bp   178 °C(lit.)
mp   80-83 °C(lit.)
solubility   alcohol: moderately soluble
  benzene: soluble
  petroleum ether: moderately soluble

Description

Application

1,2-Di-p-tolylethane has been used in the synthesis of 1-methyl-4-(4-methylphenethyl)-2-nitrobenzene,1-methyl-4-(4-methyl-2-nitrophenethyl)-2-nitrobenzene and 1,2-bis(4-methyl-3-nitrophenyl)ethane.

General description

1,2-Di-p-tolylethane undergoes oxidation by ceric ammonium nitrate in 70% aqueous acetonitrile containing nitric acid at 80°C.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DT4380000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?