EMAIL THIS PAGE TO A FRIEND
12809 Aldrich

Copper

granulated, ≥99.8% (complexometric)

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Copper, Materials Science, Metal and Ceramic Science,
InChI Key   RYGMFSIKBFXOCR-UHFFFAOYSA-N
assay   ≥99.8% (complexometric)
quality   granulated
resistivity   1.673 μΩ-cm, 20°C
granulation   0.2-0.6 mm
bp   2567 °C(lit.)
mp   1083.4 °C(lit.)
density   8.94 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves

Cross-Coupling Guide
Safety & Documentation
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?