EMAIL THIS PAGE TO A FRIEND
129496 Aldrich

Isobutyl acetate

99%

Synonym: Acetic acid isobutyl ester

  • CAS Number 110-19-0

  • Linear Formula CH3COOCH2CH(CH3)2

  • Molecular Weight 116.16

  •  Beilstein Registry Number 1741909

  •  EC Number 203-745-1

  •  MDL number MFCD00008932

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

vapor density   >4 (vs air)
vapor pressure   15 mmHg ( 20 °C)
InChI Key   GJRQTCIYDGXPES-UHFFFAOYSA-N
assay   99%
autoignition temp.   793 °F
expl. lim.   10.5 %
refractive index   n20/D 1.39(lit.)
bp   115-117 °C(lit.)
mp   −99 °C(lit.)
density   0.867 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN 1213 3 / PGII
WGK Germany 
1
RTECS 
AI4025000
Flash Point(F) 
71.6 °F
Flash Point(C) 
22 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?