• USA Home
  • 130478 - Ethyl benzoylacetate

EMAIL THIS PAGE TO A FRIEND
130478 Aldrich

Ethyl benzoylacetate

97%

Synonym: Benzoylacetic acid ethyl ester, Ethyl 3-oxo-3-phenylpropionate

  • CAS Number 94-02-0

  • Linear Formula C6H5COCH2COOC2H5

  • Molecular Weight 192.21

  •  Beilstein Registry Number 389944

  •  MDL number MFCD00009196

  • Popular Documents:  RAMAN FTIR (PDF)  

Purchase

Properties

vapor density   6.6 (vs air)
InChI Key   GKKZMYDNDDMXSE-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.52(lit.)
bp   265-270 °C(lit.)
density   1.11 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?