EMAIL THIS PAGE TO A FRIEND
131768 Aldrich

Salicylanilide

98%

Synonym: 2-Hydroxy-N-phenylbenzamide, 2-Hydroxybenzanilide, N-Phenylsalicylamide

Purchase

Properties

Related Categories Amides, Building Blocks, C8 to C20+, Carbonyl Compounds, Chemical Synthesis,
InChI Key   WKEDVNSFRWHDNR-UHFFFAOYSA-N
assay   98%
mp   136-138 °C(lit.)
solubility   H2O: slightly soluble
  H2O: slightly soluble
  alcohol: freely soluble
  benzene: freely soluble
  chloroform: freely soluble
  diethyl ether: freely soluble

Description

Packaging

100 g in poly bottle

General description

Salicylanilide inhibited mycobacterial isocitrate lyase (ICL) and showed a significant antimycobacterial effect.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
VN7850000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?