• USA Home
  • 133388 - 1,12-Dibromododecane

EMAIL THIS PAGE TO A FRIEND
133388 Aldrich

1,12-Dibromododecane

technical grade

Synonym: Dodecamethylene dibromide

Purchase

Properties

grade   technical grade
InChI Key   ZJJATABWMGVVRZ-UHFFFAOYSA-N
impurities   ≤10% 12-bromo-1-dodecanol
bp   215 °C/15 mmHg(lit.)
mp   38-42 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
JR2295000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?