EMAIL THIS PAGE TO A FRIEND
133817 Aldrich

Didecyl phthalate

  • CAS Number 84-77-5

  • Linear Formula C6H4[COO(CH2)9CH3]2

  • Molecular Weight 446.66

  •  Beilstein Registry Number 1893077

  •  MDL number MFCD00041915

  •  PubChem Substance ID 329750157

Purchase

Properties

InChI Key   PGIBJVOPLXHHGS-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?