EMAIL THIS PAGE TO A FRIEND
134872 Aldrich

Glycerol

ReagentPlus®, ≥99%

Synonym: 1,2,3-Propanetriol, Glycerin

  • CAS Number 56-81-5

  • Linear Formula HOCH2CH(OH)CH2OH

  • Molecular Weight 92.09

  •  Beilstein Registry Number 635685

  •  EC Number 200-289-5

  •  MDL number MFCD00004722

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

grade   ReagentPlus®
vapor density   3.1 (vs air)
vapor pressure   <1 mmHg ( 20 °C)
InChI Key   PEDCQBHIVMGVHV-UHFFFAOYSA-N
assay   ≥99%
autoignition temp.   698 °F
refractive index   n20/D 1.474(lit.)
bp   182 °C/20 mmHg(lit.)
mp   20 °C(lit.)
solubility   water: soluble
density   1.25 g/mL(lit.)

Description

Application

Glycerol is used both in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel’s sample well. Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component.

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MA8050000
Flash Point(F) 
320 °F
Flash Point(C) 
160 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?