• USA Home
  • 13587 - 4-Nitrophenyl carbonate Merrifield resin

EMAIL THIS PAGE TO A FRIEND
13587 Aldrich

4-Nitrophenyl carbonate Merrifield resin

extent of labeling: ~0.8 mmol/g N loading

Synonym: Activated carbonate polymer-bound, Benzyl 4-nitrophenyl carbonate polymer-bound

Purchase

Properties

Related Categories Chemical Synthesis, Other Resins, Solid Supported Synthesis, Specialty Synthesis
extent of labeling   ~0.8 mmol/g N loading
matrix   polystyrene, crosslinked with 1% DVB
particle size   100-200 mesh
storage temp.   2-8°C

Description

Other Notes

Reactive resin used to prepare immobilized carbamates from amines

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?