• USA Home
  • 13732 - (2S,3R)-(−)-3-(Benzyloxymethyl)oxirane-2-methanol 4-nitrobenzoic acid ester

EMAIL THIS PAGE TO A FRIEND
13732 Aldrich

(2S,3R)-(−)-3-(Benzyloxymethyl)oxirane-2-methanol 4-nitrobenzoic acid ester

≥99.0% (sum of enantiomers, HPLC)

Synonym: (2S,3R)-(−)-4-Benzyloxy-2,3-epoxy-1-butyl 4-nitrobenzoate

  • CAS Number 78513-08-3

  • Empirical Formula (Hill Notation) C18H17NO6

  • Molecular Weight 343.33

  •  Beilstein Registry Number 5383514

  •  PubChem Substance ID 24848321

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Epoxides, Organic Building Blocks More...
InChI Key   XBAOYRZETQXAAE-SJORKVTESA-N
assay   ≥99.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D −34±1°, c = 0.5% in chloroform
mp   78-80 °C

Description

Packaging

5 g in glass bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?