EMAIL THIS PAGE TO A FRIEND
137545 Aldrich

Tridecanenitrile

98%

  • CAS Number 629-60-7

  • Empirical Formula (Hill Notation) C13H25N

  • Molecular Weight 195.34

  •  MDL number MFCD00039492

  •  PubChem Substance ID 329750327

Purchase

Properties

InChI Key   WKJHMKQSIBMURP-UHFFFAOYSA-N
assay   98%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?