• USA Home
  • 143332 - 5-Chloro-2-hydroxy-4-methylbenzophenone

EMAIL THIS PAGE TO A FRIEND
143332 Aldrich

5-Chloro-2-hydroxy-4-methylbenzophenone

99%

Purchase

Properties

Related Categories Materials Science, Polymer Additives, Polymer Science, Stabilizers
InChI Key   IOGQACQRFGDUPT-UHFFFAOYSA-N
assay   99%
mp   138-141 °C(lit.)

Description

Packaging

10 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?