• USA Home
  • 14571 - α,ω-Bis{2-[(3-carboxy-1-oxopropyl)amino]ethyl}polyethylene glycol

EMAIL THIS PAGE TO A FRIEND
14571 Aldrich

α,ω-Bis{2-[(3-carboxy-1-oxopropyl)amino]ethyl}polyethylene glycol

Mr 10000

Synonym: O,O′-Bis[2-(succinylamino)ethyl]polyethylene glycol

Purchase

Properties

Related Categories Carboxylic Acid, Homobifunctional PEGs, Materials Science, Poly(ethylene glycol) and Poly(ethylene oxide), Polymer Science,
Quality Level   GMP
mol wt   Mr 10000
Ω-end   COOH
α-end   COOH

Price and Availability


Webinar: Recent Developments in RAFT Agents

The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Peptide PEGylation

Circulatory half-life is a key success factor of new drugs. In this respect, modification of potential candidates ranging from non-peptidic molecules to peptides and proteins with polyethyleneglycol ...
Matthias Junkers
Aldrich ChemFiles 2007, 7.2, 15.
Keywords: Degradations, Drug discovery, Eliminations, Help, Reductions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?