CAS Number 29149-93-7
Linear Formula (CH3CH2)2PCH2CH2CH2P(CH2CH3)2
Molecular Weight 220.27
Beilstein Registry Number 4951247
MDL number MFCD00233829
PubChem Substance ID 24848815
| Related Categories | Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Polydentate Phosphine Ligands |
| grade | purum |
| assay | ≥95.0% (GC) |
Bottomless glass bottle. Contents are inside inserted fused cone.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?