• USA Home
  • 14740 - N,N,N′,N′-Tetramethyl-1,3-butanediamine

EMAIL THIS PAGE TO A FRIEND
14740 Aldrich

N,N,N′,N′-Tetramethyl-1,3-butanediamine

≥99.0%

Synonym: 1,3-Bis(dimethylamino)butane, N,N,N,N′-Tetramethyl-1,3-diaminobutane

  • CAS Number 97-84-7

  • Linear Formula (CH3)2NCH(CH3)CH2CH2N(CH3)2

  • Molecular Weight 144.26

  •  Beilstein Registry Number 1698054

  •  EC Number 202-610-4

  •  MDL number MFCD00025678

  •  PubChem Substance ID 57647144

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
vapor density   5 (vs air)
vapor pressure   1.64 mmHg ( 20 °C)
InChI Key   AXFVIWBTKYFOCY-UHFFFAOYSA-N
assay   ≥99.0%
expl. lim.   7.8 %
refractive index   n20/D 1.431(lit.)
  n20/D 1.431
bp   165 °C(lit.)
  55-56 °C/12 mmHg(lit.)
density   0.787 g/mL at 25 °C(lit.)

Description

General description

N,N,N′,N′-Tetramethyl-1,3-butanediamine is an efficient electrolyte additive during the separation of tricyclic antidepressant by capillary zone electrophoresis.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2929 3(6.1) / PGII
WGK Germany 
3
RTECS 
EJ7525000
Flash Point(F) 
105.8 °F
Flash Point(C) 
41 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?