EMAIL THIS PAGE TO A FRIEND
147958 Aldrich

Pyrogallol

98%

Synonym: 1,2,3-Trihydroxybenzene

Purchase

Properties

vapor density   4.4 (vs air)
vapor pressure   10 mmHg ( 167.7 °C)
InChI Key   WQGWDDDVZFFDIG-UHFFFAOYSA-N
assay   98%
bp   309 °C(lit.)
  309 °C
mp   133-134 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1/PG 3
WGK Germany 
2
RTECS 
UX2800000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P0381 Pyrogallol, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?