EMAIL THIS PAGE TO A FRIEND
148571 Aldrich

Acrylamide

97%

Synonym: 2-Propenamide, Acrylic acid amide

Purchase

Properties

vapor density   2.45 (vs air)
vapor pressure   0.03 mmHg ( 40 °C)
InChI Key   HRPVXLWXLXDGHG-UHFFFAOYSA-N
assay   97%
contains   25-30 ppm cupric ion as stabilizer
bp   125 °C/25 mmHg(lit.)
mp   82-86 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 2074 6.1 / PGIII
WGK Germany 
3
RTECS 
AS3325000
Flash Point(F) 
280.4 °F
Flash Point(C) 
138 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A8887 Acrylamide, suitable for electrophoresis, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?