• USA Home
  • 14971 - 3-(Boc-aminomethyl)benzoic acid

EMAIL THIS PAGE TO A FRIEND
14971 Aldrich

3-(Boc-aminomethyl)benzoic acid

≥97.0% (HPLC)

  • CAS Number 117445-22-4

  • Linear Formula (CH3)3CO2CNHCH2C6H4CO2H

  • Molecular Weight 251.28

  •  Beilstein Registry Number 6957774

  •  MDL number MFCD00273426

  •  PubChem Substance ID 57647232

Purchase

Properties

Related Categories Aromatic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   MQNHKLMHRZYTBZ-UHFFFAOYSA-N
assay   ≥97.0% (HPLC)

Description

Packaging

1, 5 g in poly bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?