• USA Home
  • 14977 - Boc-β-Homoglu(OBzl)-OH

EMAIL THIS PAGE TO A FRIEND
14977 Aldrich

Boc-β-Homoglu(OBzl)-OH

≥98.0% (TLC)

Synonym: (S)-3-(Boc-amino)adipic acid 6-benzyl ester, Boc-L-β-homoglutamic acid 6-benzyl ester

  • CAS Number 218943-30-7

  • Empirical Formula (Hill Notation) C18H25NO6

  • Molecular Weight 351.39

  •  Beilstein Registry Number 8079168

  •  MDL number MFCD01862936

  •  PubChem Substance ID 24848968

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   JABNOCWCLLYHKE-AWEZNQCLSA-N
assay   ≥98.0% (TLC)

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?