EMAIL THIS PAGE TO A FRIEND
14984 Aldrich

Boc-Phe(2-CN)-OH

≥98.0% (TLC)

Synonym: Boc-2-cyano-L-phenylalanine

  • CAS Number 216312-53-7

  • Empirical Formula (Hill Notation) C15H18N2O4

  • Molecular Weight 290.31

  •  Beilstein Registry Number 9410479

  •  MDL number MFCD01860623

  •  PubChem Substance ID 24848977

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   AJHPGXZOIAYYDW-LBPRGKRZSA-N
assay   ≥98.0% (TLC)

Description

Packaging

1 g in poly tube

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?