EMAIL THIS PAGE TO A FRIEND
15011 Aldrich

Boc-Phe(2-CF3)-OH

≥98.0% (TLC)

Synonym: Boc-2-(trifluoromethyl)-L-phenylalanine

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   XKMOOODKNPYTEE-NSHDSACASA-N
assay   ≥98.0% (TLC)

Description

Packaging

1 g in poly tube

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?