EMAIL THIS PAGE TO A FRIEND
15016 Aldrich

Boc-D-Phe(4-CF3)-OH

≥98.0% (TLC)

Synonym: Boc-4-(trifluoromethyl)-D-phenylalanine

  • CAS Number 82317-83-7

  • Empirical Formula (Hill Notation) C15H18F3NO4

  • Molecular Weight 333.30

  •  Beilstein Registry Number 5125776

  •  MDL number MFCD00797557

  •  PubChem Substance ID 57647201

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   SMVCCWNHCHCWAZ-LLVKDONJSA-N
assay   ≥98.0% (TLC)
optical activity   [α]20/D −5.4±0.7°, c = 1% in methanol

Description

Packaging

1 g in poly bottle

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?