EMAIL THIS PAGE TO A FRIEND
15023 Aldrich

Boc-D-Phe(2-F)-OH

≥98.0% (TLC)

Synonym: Boc-2-fluoro-D-phenylalanine

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   NTWUXBKUDXGMHV-LLVKDONJSA-N
assay   ≥98.0% (TLC)
optical activity   [α]20/D −10±1°, c = 1% in ethyl acetate
mp   94-98 °C

Description

Packaging

1 g in poly tube

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?