• USA Home
  • 15039 - Boc-β-(3-benzothienyl)-D-Ala-OH

EMAIL THIS PAGE TO A FRIEND
15039 Aldrich

Boc-β-(3-benzothienyl)-D-Ala-OH

≥97.0% (HPLC)

Synonym: Boc-3-(3-benzothienyl)-D-alanine

  • CAS Number 111082-76-9

  • Empirical Formula (Hill Notation) C16H19NO4S

  • Molecular Weight 321.39

  •  Beilstein Registry Number 8995535

  •  MDL number MFCD00079679

  •  PubChem Substance ID 24849025

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   MURVSBJYXHTRJQ-GFCCVEGCSA-N
assay   ≥97.0% (HPLC)

Description

Packaging

1 g in poly tube

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?