EMAIL THIS PAGE TO A FRIEND
15077 Aldrich

Boc-D-His(Bom)-OH

≥98.0% (TLC)

Synonym: Nα-Boc-π-Bom-D-histidine, Nα-Boc-N(im)-benzyloxymethyl-D-histidine

  • CAS Number 99310-01-7

  • Empirical Formula (Hill Notation) C19H25N3O5

  • Molecular Weight 375.42

  •  MDL number MFCD00076940

  •  PubChem Substance ID 57647296

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Histidine, Peptide Chemistry More...
InChI Key   LPVKZCHCZSFTOJ-MRXNPFEDSA-N
assay   ≥98.0% (TLC)

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?