EMAIL THIS PAGE TO A FRIEND
15191 Aldrich

Boc-D-Val-OH

≥98.0% (TLC)

Synonym: Boc-D-valine

  • CAS Number 22838-58-0

  • Empirical Formula (Hill Notation) C10H19NO4

  • Molecular Weight 217.26

  •  Beilstein Registry Number 2050408

  •  MDL number MFCD00038282

  •  PubChem Substance ID 57647339

  •  eCl@ss 32160406

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Valine Amino Acids More...
InChI Key   SZXBQTSZISFIAO-SSDOTTSWSA-N
assay   ≥98.0% (TLC)

Description

Packaging

5 g in poly bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?