EMAIL THIS PAGE TO A FRIEND
15347 Aldrich

Boc-1-Nal-OH

≥97.0% (HPLC)

Synonym: Boc-3-(1-naphthyl)-L-alanine

  • CAS Number 55447-00-2

  • Empirical Formula (Hill Notation) C18H21NO4

  • Molecular Weight 315.36

  •  Beilstein Registry Number 3561332

  •  MDL number MFCD00085377

  •  PubChem Substance ID 24849345

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   KHHIGWRTNILXLL-HNNXBMFYSA-N
assay   ≥97.0% (HPLC)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?