• USA Home
  • 15447 - Boc-N-Me-Phe-OH (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
15447 Aldrich

Boc-N-Me-Phe-OH (dicyclohexylammonium) salt

≥97.0% (CHN)

Synonym: Boc-N-methyl-L-phenylalanine (dicyclohexylammonium) salt

  • CAS Number 40163-88-0

  • Linear Formula C6H5CH2CH(COOH)N(CH3)COOC(CH3)3 · C12H23N

  • Molecular Weight 460.65

  •  Beilstein Registry Number 6310237

  •  MDL number MFCD00070397

  •  PubChem Substance ID 24849445

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine More...
InChI Key   MFORGORUOHHLHW-YDALLXLXSA-N
assay   ≥97.0% (CHN)
optical activity   [α]20/D −23.5±1°, c = 1% in methanol
mp   174-177 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?