EMAIL THIS PAGE TO A FRIEND
15453 Aldrich

H-Lys(Boc)-OH

purum, ≥97.0% (TLC)

Synonym: Nε-Boc-L-lysine

  • CAS Number 2418-95-3

  • Linear Formula (CH3)3COCONH(CH2)4CH(NH2)COOH

  • Molecular Weight 246.30

  •  Beilstein Registry Number 2417626

  •  MDL number MFCD00037221

  •  PubChem Substance ID 24849451

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Lysine, Peptide Chemistry More...
InChI Key   VVQIIIAZJXTLRE-QMMMGPOBSA-N
grade   purum
assay   ≥97.0% (TLC)
optical activity   [α]20/D +17±1°, c = 1% in acetic acid
mp   250 °C (dec.)(lit.)
  ~250 °C (dec.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?