EMAIL THIS PAGE TO A FRIEND
15474 Aldrich

Boc-Cit-OH

technical, ≥84% (T)

Synonym: Nα-Boc-L-citrulline

  • CAS Number 45234-13-7

  • Empirical Formula (Hill Notation) C11H21N3O5

  • Molecular Weight 275.30

  •  Beilstein Registry Number 6335956

  •  MDL number MFCD00080271

  •  PubChem Substance ID 57647449

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Glutamine Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   CMJCWQISQGNHHI-ZETCQYMHSA-N
grade   technical
assay   ≥84% (T)
optical activity   [α]20/D −8.7±1°, c = 1% in H2O
storage temp.   2-8°C

Description

Packaging

5 g in glass bottle

Price and Availability


Laboratory Gloves

Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?