EMAIL THIS PAGE TO A FRIEND
15504 Aldrich

Boc-Tic-OH

≥97.0% (TLC)

Synonym: (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • CAS Number 78879-20-6

  • Empirical Formula (Hill Notation) C15H19NO4

  • Molecular Weight 277.32

  •  Beilstein Registry Number 4297114

  •  MDL number MFCD00143845

  •  PubChem Substance ID 24849331

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Other Unnatural Amino Acids & Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   HFPVZPNLMJDJFB-LBPRGKRZSA-N
assay   ≥97.0% (TLC)
optical activity   [α]20/D +19.0±1.5°, c = 0.7% in methanol
storage temp.   2-8°C

Description

Other Notes

Methylene-bridged derivative of BOC-L-phenylalanine.

Packaging

1 g in poly bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?