• USA Home
  • 15909 - Trihexyltetradecylphosphonium tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
15909 Aldrich

Trihexyltetradecylphosphonium tetrafluoroborate

Synonym: Tetradecyltrihexylphosphonium tetrafluoroborate

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

CYPHOS® Phosphonium Ionic Liquids

The phosphonium cation, with the generic formula [PR3R’]+ and with a judicious selection of the appropriate anion, forms many phosphonium salts that are liquid at room temperature and many have melti...
Chemfiles Volume 5 Article 6
Keywords: Industries, Melting, Solvents

ChemFiles Volume 4 Article 9 - Ionic Liquids

Ionic Liquids have experienced a comet-like boost in the last few years. This fascinating class of new materials with its unusual and extraordinary properties has found innumerable new applications i...
ChemFiles Volume 4 Article 9
Keywords: Addition reactions, Applications, Industries, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?