EMAIL THIS PAGE TO A FRIEND
16011 Aldrich

Brij® O10

main component: decaethylene glycol oleyl ether

Synonym: Polyethylene glycol oleyl ether

Purchase

Description

Legal Information

Brij is a registered trademark of Croda International PLC

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
RK2800000
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?