• USA Home
  • 160202 - N,N,N′,N′-Tetramethyl-p-phenylenediamine

EMAIL THIS PAGE TO A FRIEND
160202 Aldrich

N,N,N′,N′-Tetramethyl-p-phenylenediamine

98%

Synonym: TMPD, TMPDA, TMPPD, Wurster’s reagent

Purchase

Properties

InChI Key   CJAOGUFAAWZWNI-UHFFFAOYSA-N
assay   98%
bp   260 °C(lit.)
mp   49-51 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
1
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

T7394 N,N,N′,N′-Tetramethyl-p-phenylenediamine, 99%, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?