• USA Home
  • 16055 - Bromine on polymer support

EMAIL THIS PAGE TO A FRIEND
16055 Aldrich

Bromine on polymer support

extent of labeling: 1.2-1.8 mmol/g active bromine (Br3-) loading

Synonym: Amberlite® IRA-900 Br3--form

Purchase

Properties

Related Categories Bromination, C-X Bond Formation, C-X Bond Formation (Halogen), Chemical Synthesis, Solid Supported Synthesis,
extent of labeling   1.2-1.8 mmol/g active bromine (Br3-) loading
impurities   ~0.03 mmol/g Cl- (resin)

Description

Other Notes

Polymer supported brominating agent

Packaging

5, 25 g in glass bottle

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3260 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?