EMAIL THIS PAGE TO A FRIEND
161071 Aldrich

1,2-Naphthoquinone

technical grade

Synonym: β-Naphthoquinone

Purchase

Properties

grade   technical grade
InChI Key   KETQAJRQOHHATG-UHFFFAOYSA-N
mp   139-142 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
QL7000000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?